Learn More
Akro-Mils™ Akro-Grid Storage Totes: 1.66 cu. Ft./50 lb.
With optional lengthwise and crosswise dividers for smaller storage
Manufacturer:  Akro-Mils™ 33220RED
 View more versions of this product
Catalog No. 08-757-867
Add to cart
Description
Description
These lightweight totes facilitate work-in-progress transfers; enable compartmentalized storage for items used in concert.
  • Offer hundreds of storage configurations; can be subdivided down to a 1 1/8 in. square
  • Injection-molded from polypropylene block copolymer
  • Resist most solvents and chemicals
  • Choice of Red, Blue or Gray for color-coding and easy identification
  • Available in eight sizes, many with the same footprint for secure stacking
  • Stacking rims and external ribbing add strength
  • Compatible with most racks, shelving and automated material handling equipment
  • Dividers, lids and label holder sold separately
Specifications
Specifications
22.38 x 17.38 x 10 in. (56.8 x 44 x 25.4cm)
1.66 cu. ft.
25.4cm
56.8cm
44cm
19.9 in.
15.25 in.
20.12 in.
47L
10 in.
22.38 in.
17.38 in.
Red
50.5cm
38.7cm
51cm