Learn More
Akro-Mils™ InSight™ Storage System: 10 lb., 53/8 x 4/18 x 31/4 in.
Visible organization solutions for healthcare and industrial workspaces
Manufacturer:  Akro-Mils™ 405A1
 View more versions of this product
Catalog No. 08-757-670
Add to cart
Description
Description
InSight bins offer transparent storage with a sleek, modern design. Can be stacked, shelved or hung on Akro-Mils rails and louvered panels or ReadySpace™ Storage System.
  • Constructed of durable polycarbonate
  • Autoclavable
  • No tip feature for secure stacking
  • Rear lip supports bin on hanging systems
  • Large label area for easy identification and bar code scanning
  • Optional lids and length/width dividers available to create dust-free, custom-size compartments
Specifications
Specifications
Yes
Length divider
Polycarbonate
InSight Storage System