Learn More
Akron Brass™ Adjustable Hydrant Wrenches Non-distribution product as customer accommodation.
Manufacturer:  Akron Brass™ 17
Catalog No. 19-805-805
Please call customer service for current pricing. 1-800-766-7000
Description
Description
  • Adjustable to 1 3/4 in. pentagon or 1 1/4 in. square nut
  • Plated for corrosion resistance
  • Designed to prevent hydrant nut damage
  • Spanner fits 2 1/2 in. to 4 1/2 in. rocker lug or pin lug couplings
  • Replaceable brass inserts
Specifications
Specifications
Adjustable hydrant wrench
2.5 to 4.5 in. rocker lug or pin lug couplings
47.29cm
4.5 lb.
Prevent hydrant nut damage
18.62 in.
2kg