Learn More
Akron Brass™ Coast Guard Shipboard Vari-Nozzles Non-distribution product as customer accommodation.
Manufacturer:  Akron Brass™ 3022
Catalog No. 19-121-816
Please call customer service for current pricing. 1-800-766-7000
Description
Description
  • Brass construction
  • Low friction ball and dual seats for smooth operation
  • Molded teeth
  • Rubber-like bumper and handle
  • Constant gallonage in fog and straight stream patterns
  • Flush without shutting down
Specifications
Specifications
8.62 x 4.25 x 6 in. (21.9 x 10.79 x 15.24cm)
6 in.
Low friction ball, dual seats, molded teeth, Rubber-like bumper
8.62 in.
100psi
3.2kg
10.79cm
Vari nozzle
360
15.24cm
1.5 in.
21.9cm
7.12 lb.
4.25 in.
Brass