Learn More
Akron Brass In-Line Eductor
Manufacturer:  AKRON BRASS CO. 3060
 View more versions of this product
Catalog No. 19-823-710
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
60 gpm
1.5/2.5 in. swivel
19.7cm
3.5 lb.
Pyrolite™
Foam equipment
7.75 in.
200psi
1.6kg
Pyrolite In-Line Foam Eductor