Learn More
Akron Brass™ In-Line Eductor Non-distribution product as customer accommodation.
Manufacturer:  Akron Brass™ 2325
 View more versions of this product
Catalog No. 19-805-815
Add to cart
Description
Description
  • Large, easy to read, and removable metering dial with infinite settings between 0% and 6%
  • The Eductors are rated to flow at 200 psi (14 bar) inlet pressure and work effectively with nozzles rated from 50 to 100 psi (3 to 7 bar)
  • 1.5 in. (38 mm) or 2.5 in. (65 mm) swivel inlet x 1.5 in. (38mm) male
  • 30 in. pickup hose standard on 60 gpm (230 lpm) eductors, 48 in. hose optional
Specifications
Specifications
95 gpm
1.5 in. swivel
25.7cm
7 lb.
N/A
Foam equipment
10.25 in.
150psi
3.1kg
In-Line Foam Eductor