Learn More
Akron Brass™ Spanner and Hydrant Wrenches Non-distribution product as customer accommodation.
Two wrenches available for a variety of applications
Specifications
For Use With (Application) Prevent hydrant nut damage
View More Specs
Products 1
Catalog Number Mfr. No. Type For Use With (Equipment) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Type For Use With (Equipment) Length (Metric) Price Quantity & Availability  
19-805-803 Non-distribution product as customer accommodation.
View Documents
Akron Brass™
15/DUP
Hydrant and Spanner Wrench 2.5 to 4.5 in. rocker lug or pin lug couplings 49.53 cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
Universal Spanner Wrench
  • All-purpose wrench for 0.75 in. to 3 in. rocker lug and pin lug couplings features belt hook eye, hammer head and gas cock shutoff
  • With Akrolite™ construction
Hydrant and Spanner Wrench
  • Designed to help prevent damage to hydrant nuts
  • Adjusts to a pentagon nut up to 1.75 in. (measured from point to flat) or to a square nut up to 1.25 in.
  • Spanner fits 2.5 in. to 4.5 in. rocker lug or pin lug couplings
  • Plated for corrosion resistance
  • With replaceable brass insert
Specifications
Specifications
Prevent hydrant nut damage