Learn More
Akron Brass™ Spanner and Hydrant Wrenches Available on GSA/VA Contract for Federal Government customers only.
Two wrenches available for a variety of applications
Manufacturer:  AKRON BRASS CO. 10
 View more versions of this product
Catalog No. 19-805-798
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Firefighting, wildland forestry
11.5 in.
29.21cm
0.22kg
0.75 to 3 in. rocker lug and pin lug couplings
Universal spanner wrench
Akrolite
0.5 lb.