Learn More
Akron Brass™ Spanner and Hydrant Wrenches Available on GSA/VA Contract for Federal Government customers only.
Two wrenches available for a variety of applications
Manufacturer:  AKRON BRASS CO. 15
 View more versions of this product
Catalog No. 19-805-803
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Prevent hydrant nut damage
19.5 in.
49.53cm
2.26kg
2.5 to 4.5 in. rocker lug or pin lug couplings
Hydrant and spanner wrench
Plated
5 lb.