Learn More
Akron Brass™ Turbojet Nozzles Non-distribution product as customer accommodation.
Manufacturer:  Akron Brass™ 1702
 View more versions of this product
Catalog No. 19-805-807
Please call customer service for current pricing. 1-800-766-7000
Description
Description
  • Pattern Identification
  • Larger more legible labels
  • Simple to operate (1/4 turn from straight stream to fog)
  • Raised lug marks straight stream for limited visibility operations
  • Easy to service & repair
  • Meets European Standard
  • Flow setting at 100 psi (7 bar) operating pressure can be used for high pressure applications up to 580 psi (40 bar) hydrostatic pressure
  • Raised bumper lug for straight stream pattern identification
  • Compatible with Quick-Attack Foam Tube Style 755
  • SOLAS & MarEd approved available (Must specify)
Specifications
Specifications
NFPA 1964
Quick attack Foam tube style 755
1 in.
20.6cm
3 lb.
Pyrolite™
50 to 100 and 150 to 230
Pistol grip
9.25 in.
100psi
1.4kg
Turbojet™ nozzle