Learn More
Akron Turbojet Nozzles
Manufacturer:  AKRON BRASS CO. 1727
 View more versions of this product
Catalog No. 19-805-811
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1964
Quick attack Foam tube style 755
1.5 in.
17.8cm
4.5 lb.
Pyrolite™
475 to 550 and 750 to 950
Twist shut-off
7 in.
100psi
2kg
High range Turbojet™ nozzle