Learn More
Medchemexpress LLC HY-18296 5mg Medchemexpress, AKT-IN-1 CAS:1357158-81-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY182965MG
Encompass_Preferred
Medchemexpress, HY-18296 5mg AKT-IN-1 CAS:1357158-81-6 AKT-IN-1 is an allosteric AKT inhibitor with an IC50 of 1.042 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0004
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.