Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0647-2L45, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236780AF532

Catalog No. NP236780A32


Add to cart

Description

Description

AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

AKT
MM0647-2L45
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human AKT1).
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human AKT1, 2 and 3 by WB. Cross reacts with mouse and rat AKTs proteins.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG2
0.1 mL
Primary
207.0
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit