Learn More
AKT Mouse anti-Human, DyLight 550, Clone: MM0647-2L45, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236780R
Catalog No. NBP236780R
Add to cart
Description
Description
AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AKT
MM0647-2L45
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human AKT1).
Protein G purified
Store at 4C in the dark.
Monoclonal
Human AKT1, 2 and 3 by WB. Cross reacts with mouse and rat AKTs proteins.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG2
0.1 ml
Primary
207
Human