Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT (p Ser473) Mouse anti-Human, Biotin, Clone: MM0648-7L25, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236781B

Catalog No. NBP236781B


Add to cart

Description

Description

AKT p Ser473 Monoclonal antibody specifically detects AKT p Ser473 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AKT (p Ser473)
MM0648-7L25
Western Blot
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with phosphopeptide containing human AKT1 (phosphor-S473).
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosph-AKT1, 2 and 3 at S472, S473, and S474 WB. It cross reacts with mouse phosphor AKT proteins.
Western Blot
Biotin
PBS with 0.05% Sodium Azide
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG1
0.1 ml
Primary
207
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit