Learn More
AKT (p Ser473) Mouse anti-Human, HRP, Clone: MM0648-7L25, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236781H
Catalog No. NBP236781H
Add to cart
Description
Description
AKT p Ser473 Monoclonal antibody specifically detects AKT p Ser473 in Human samples. It is validated for Western Blot.
Specifications
Specifications
AKT (p Ser473)
MM0648-7L25
Western Blot
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with phosphopeptide containing human AKT1 (phosphor-S473).
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosph-AKT1, 2 and 3 at S472, S473, and S474 WB. It cross reacts with mouse phosphor AKT proteins.
Western Blot
HRP
PBS with No Preservative
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG1
0.1 ml
Primary
207
Human