Learn More
AKT (p Thr308) Mouse anti-Human, DyLight 755, Clone: MM0650-9F42, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236782IR
Catalog No. NBP236782IR
Add to cart
Description
Description
AKT p Thr308 Monoclonal antibody specifically detects AKT p Thr308 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AKT (p Thr308)
MM0650-9F42
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing the human Akt1 T308 site.
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes human AKT1 with phospho-T308.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG3
0.1 ml
Primary
207
Human