Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP303025100UL
Description
AKT1 Polyclonal antibody specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, KnockDownSpecifications
AKT1 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100 μL | |
Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases | |
Primary | |
207 |
Immunohistochemistry, KnockDown, Western Blot | |
PBS with 50% glycerol, pH7.3. | |
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1 | |
A synthetic peptide of human AKT1. | |
Affinity purified | |
RUO | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title