Learn More
Enzo Life SciencesSupplier Diversity Partner Akt1/2 (pSer473/474) ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900162
The [pSer473/474]Akt1/2 EIA kit is a colorimetric immunometric enzyme immunoassay kit. Absorbance is read at 450 nm. This is an ultra-sensitive assay that lets you measure as little as 5.5 pg/mL Akt1 or Akt2 phosphorylated on Ser473 or Ser474. Obtain fully quantitative results from up to 41 samples in duplicate in just 3 hours. The assay has been validated with up and down regulation experiments and compared to Western blot. Alternative Name: PKB. Protein kinase B. Sensitivity: 5.5 pg/ml (range 17.5 - 560 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 32, Wash buffer concentrate, Standard, TMB Substrate, Cell lysis buffer 5, Stop solution 2
Catalog No. 50-658-188