Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI4D6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504230

Catalog No. 50-167-4958


Add to cart

Description

Description

The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.
Specifications

Specifications

AKT1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P31749, P31750, P47196
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11651, 207, 24185, 490878
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4D6
Unconjugated
AKT1
Liquid
AKT1
Full length human recombit protein of human AKT1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit