Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Mouse anti-Human, Mouse, Clone: 6D5, Novus Biologicals™

Mouse Monoclonal Antibody

$204.00 - $308.00

Specifications

Antigen AKT1
Clone 6D5
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry 2-5 ug, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60246100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP260246100UL
100 ul Each for $308.00
Add to cart
 
NB6024650UL View Documents Novus BiologicalsSupplier Diversity Partner
NBP26024650UL
50 ul Each for $204.00
Add to cart
 
Description

Description

AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

AKT1
1 mg/mL
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse
AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1
AKT1
IgG2a Kappa
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
6D5
Western Blot 1:1000, Flow Cytometry 2-5 ug, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Unconjugated
Mouse
Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Signal Transduction
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
207
Recombinant human AKT1 (1-480aa) purified from insect cell.
Monoclonal
Protein A purified

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit