Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen AKT1
Applications Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092898
View Documents
Novus Biologicals
NBP303025100UL
100 μg Each for $320.71
Add to cart
 
NB092899
View Documents
Novus Biologicals
NBP30302520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

AKT1 Polyclonal antibody specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, KnockDown
Specifications

Specifications

AKT1
Unconjugated
Rabbit
Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases
PBS with 50% glycerol, pH7.3.
207
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1
A synthetic peptide of human AKT1.
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit