Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1, Rabbit anti-Human, SureLight 488, Clone: G12, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18789

Catalog No. 89-335-623


Add to cart

Specifications

Specifications

AKT1
G12
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
v-akt murine thymoma viral oncogene homolog 1
AKT|MGC99656|PKB|PKB-ALPHA|PRKBA|RAC|RAC-ALPHA
Rabbit
IgG1, kappa
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
SureLight 488
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Liquid
AKT1
A synthetic phosphopeptide corresponding to residues surrounding T308 of human AKT1.
100 Reactions
Store at 4°C.
Primary
207
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit