Learn More
AKT1 Rabbit anti-Human, Unconjugated, Clone: EBG-1, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB22031
Catalog No. 89-339-769
Add to cart
Description
Description
AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for IP, WB-Ce, IHC-P, IF.
Specifications
Specifications
AKT1
EBG-1
v-akt murine thymoma viral oncogene homolog 1
Liquid
Rabbit
IgG
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
AKT1
AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
A synthetic peptide corresponding to human AKT1
100 μg
Primary
207