Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4H7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270146AF647

Catalog No. N270146A647


Add to cart

Description

Description

AKT2 Monoclonal antibody specifically detects AKT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

AKT2
OTI4H7
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
AKT2
Full length recombinant protein of human AKT2 (NP_001617) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Signal Transduction
Primary
208.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit