Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT2, Rabbit anti-Human, FITC, Clone: S7-R, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB16008
Specifications
| AKT2 | |
| Monoclonal | |
| FITC | |
| In PBS (10 mg/mL BSA, 0.05% Sodium Azide). | |
| PKBB/PKBBETA/PRKBB/RAC-BETA | |
| Rabbit | |
| RUO | |
| 208 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| Liquid |
| Flow Cytometry | |
| S7-R | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| v-akt murine thymoma viral oncogene homolog 2 | |
| AKT2 | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction