Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT2, Rabbit anti-Human, FITC, Clone: Y14-H, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB16007
Specifications
AKT2 | |
Monoclonal | |
FITC | |
In PBS (10 mg/mL BSA, 0.05% Sodium Azide). | |
PKBB/PKBBETA/PRKBB/RAC-BETA | |
Rabbit | |
RUO | |
208 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
Liquid |
Flow Cytometry | |
Y14-H | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
v-akt murine thymoma viral oncogene homolog 2 | |
AKT2 | |
100 Reactions | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction