Learn More
AKT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 176091AP
Catalog No. 50-173-1681
Add to cart
Description
Description
Akt, protein kinase B (PKB), is a serine/threonine kinase, which is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.
Specifications
Specifications
AKT2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P31751, P47197, Q60823
AKT2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt 2, Protein kinase B beta, RAC BETA, RAC PK beta
Rabbit
IgG
150 μL
-20°C
Primary
11652, 208, 25233
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
AKT2
Liquid
Akt2
AKT2 Fusion Protein Ag11770
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat