Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 Mouse anti-Human, Mouse, PE/Cy5.5, Clone: OTI9B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271528PECY55

Catalog No. N271528PEC5


Add to cart

Description

Description

AKT3 Monoclonal antibody specifically detects AKT3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

AKT3
OTI9B2
Flow Cytometry
Affinity Purified, Conjugated
AKT3
Full length recombinant protein of human AKT3 (NP_005456) produced in E.coli.
Immunogen affinity purified
Cancer, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction
Primary
10000.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Mouse
IgG3
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit