Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT3 (PKB gamma) Recombinant Human Protein Non-distribution product as customer accommodation.

AKT3 (PKB gamma)

$429.00

Specifications

Accession Number NP_005456
Assays AKT3 (PKB gamma) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM Na3VO4, 1mM DTT, 0.01% Triton™ X-100, 200μM ATP, 50μM crosstide peptide substrate (GRPRTSSFAEG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name AKT3 (PKB gamma) Recombinant Human Protein
Expression System Baculovirus
Gene Alias AKT3 (PKB gamma)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3185 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3185
10μg Each for $429.00
Add to cart
 
Description

Description

AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). It is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake.
Specifications

Specifications

NP_005456
AKT3 (PKB gamma) Recombinant Human Protein
AKT3 (PKB gamma)
60.4kDa
1 Each
95% by SDS-PAGE
–80°C
Human
AKT3 (PKB gamma) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM Na3VO4, 1mM DTT, 0.01% Triton™ X-100, 200μM ATP, 50μM crosstide peptide substrate (GRPRTSSFAEG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
AKT
AKT3 (PKB gamma) Recombinant Human Protein
AKT3 (PKB gamma) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit