Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKTIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148601AP

Catalog No. 50-172-9783


Add to cart

Description

Description

Component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex). Regulates apoptosis by enhancing phosphorylation and activation of AKT1. Increases release of TNFSF6 via the AKT1/GSK3B/NFATC1 signaling cascade.
Specifications

Specifications

AKTIP
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5FVH4, Q64362, Q9H8T0
AKT interacting protein, AKTIP, FT1, FTS, Fused toes protein homolog
Rabbit
IgG
150 μL
-20°C
Primary
14339, 291906, 64400
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
AKTIP
Liquid
Aktip
AKTIP Fusion Protein Ag6662
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit