Al-23 Grinding Pieces, H: 10mm, 440 Pieces/Kg, Alfa Aesar™

Manufacturer: Alfa Aesar 33190A7

Quantity 5kg (5 x 440 Pieces)

 View more versions of this product

Catalog No. AA33190A7

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Grinding Pieces
    Chemical Name or Material Al-23 Grinding Pieces

    Specifications

    Quantity 5kg (5 x 440 Pieces)
    Physical Form Grinding Pieces
    Chemical Name or Material Al-23 Grinding Pieces

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit