Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Al-23 Multibore Round Tube, O.D.: 5.5mm, Four 1.2mm Bores, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 32712EH

 View more versions of this product

Catalog No. AA32712EH

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Al-23 Multibore Round Tube
Four bores
600 mm
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

TSCA : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit