Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Al-23 Multibore Round Tube, O.D.: 5mm, Two 1.2mm Bores, Alfa Aesar
Manufacturer: Alfa Aesar 32654EM
Specifications
Al-23 Multibore Round Tube | |
800mm | |
Two bores |
Safety and Handling
TSCA : U
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title