Al-23 Solid Rod, Round, Dia.: 8mm, Alfa Aesar™

Manufacturer: Alfa Aesar 32935DW

Quantity 300mm

 View more versions of this product

Catalog No. AA32935DW

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Solid Round Rod
    Chemical Name or Material Al-23 Solid Rod

    Specifications

    Quantity 300mm
    Physical Form Solid Round Rod
    Chemical Name or Material Al-23 Solid Rod

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit