Learn More
Sorbent Technologies IncSupplier Diversity Partner Alumina N TLC Plates, w/UV254, polyester backed, 200um, 20x20cm, 25/pkApplications: terpenes, alkaloids, steroids, aliphatic and aromatic compounds.To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120°C. DFS Item
Manufacturer:  Sorbent Technologies IncSupplier Diversity Partner 0224126
Alumina N TLC Plates, w/UV254, polyester backed, 200um, 20x20cm, 25/pkAlumina Neutral (pH 6-8) with UV254, analytical, polyester backed TLC plates. The coating thickness is 200um and dimensions are 20x20cm. There are 25 plates per pack.Applications include: terpenes, alkaloids, steroids, aliphatic and aromatic compounds.Our Alumina N TLC plates are coated with aluminum oxide (about pH 6-8). To ensure sufficient adherence an inert organic binder is added to the aqueous suspension for coating. The aluminum oxide layer has a specific surface of about 200 m2/g (BET) and a pore size of about 60 Å.To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120°C.
Catalog No. 50-113-8482