Learn More
[Ala9] Autocamtide 2, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66658EXD
 View more versions of this product
Catalog No. AAJ66658EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1497.76
0.5mg
Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu
-30° to -10°C
[Ala9] Autocamtide 2
HPLC
Solid
RUO
Autocamtide-2-Related Inhibitory Peptide