Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Alanine, ≥98%, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
200g
1kg
5kg
Packaging:
Plastic bottle
107-95-9
C3H7NO2
89.094
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine,3-aminopropionic acid,abufene,beta alanine,h-beta-ala-oh,beta-aminopropionic acid,2-carboxyethylamine,.beta.-alanine,alanine, beta,propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
C(CN)C(=O)O
107-95-9
C3H7NO2
89.094
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine,3-aminopropionic acid,abufene,beta alanine,h-beta-ala-oh,beta-aminopropionic acid,2-carboxyethylamine,.beta.-alanine,alanine, beta,propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
C(CN)C(=O)O
Linear Formula NH2CH2CH2COOH
Packaging Plastic bottle
Formula Weight 89.09
Physical Form Crystalline Powder
Percent Purity ≥98%
Melting Point ~205° (decomposition)
Quantity 5kg
Specific Gravity 1.437
Chemical Name or Material beta-Alanine