Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Alanine, ≥98%, Ultrapure, Thermo Scientific™

Catalog Number AAJ1066528
Click to view available options
Quantity:
200 g
1 kg
5 kg
Packaging:
Plastic Bottle
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽205°C (decomposition)
Linear Formula NH2CH2CH2COOH
Beilstein 906793
Packaging Plastic Bottle
Formula Weight 89.09
Percent Purity ≥98%
Density 1.437
Quantity 200 g
Specific Gravity 1.437
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine
Show More Show Less
EINECSNumber 203-536-5
RTECSNumber UA2369200
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit