Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Alanine-tert-butylester hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
58620-93-2
C7H15NO2·ClH
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride,h-beta-ala-otbu.hcl,beta-alanine tert-butyl ester hydrochloride,h-beta-ala-otbu hcl,beta-alanine t-butyl ester hydrochloride,3-amino-propionic acid tert-butyl ester hydrochloride,h-?-ala-otbu.hcl,h-,a-ala-otbu.hcl,h-beta-ala-otbu hydrochloride,tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
CC(C)(C)OC(=O)CCN.Cl
58620-93-2
C7H15NO2·ClH
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride,h-beta-ala-otbu.hcl,beta-alanine tert-butyl ester hydrochloride,h-beta-ala-otbu hcl,beta-alanine t-butyl ester hydrochloride,3-amino-propionic acid tert-butyl ester hydrochloride,h-?-ala-otbu.hcl,h-,a-ala-otbu.hcl,h-beta-ala-otbu hydrochloride,tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
CC(C)(C)OC(=O)CCN.Cl
Melting Point 167.8°C to 168.5°C
Infrared Spectrum Authentic
Color Colorless
Quantity 5g
Packaging Glass bottle
Elemental Analysis 45.1 to 47.4 % C 7.5 to 7.9 % N
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 181.66
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material β-Alanine-tert-butylester hydrochloride
Show More Show Less