Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alanine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
56-41-7
C3H7NO2
89.09
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
2-aminopropanoic acid
CC(N)C(O)=O
56-41-7
C3H7NO2
89.09
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
2-aminopropanoic acid
CC(N)C(O)=O

A1253, 56-41-7, C3H7NO2

Spectrum™ Alanine, USP is a non-essential alpha-amino acid and appears as a white powder. Alanine is used in clinical nutrition and as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Chloride 0.0005
Ignition Residue 0.0015
Iron (Fe) 30 ppm
Loss on Drying 0.002
Packaging Amber Glass Bottle
Specific Rotation +13.7° to +15.1°
Percent Purity 98.5 to 101.5%
Sulfate 0.0003
Quantity 5 g