Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alanine, USP, 98.5-101.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Alanine, USP is a non-essential alpha-amino acid and appears as a white powder. Alanine is used in clinical nutrition and as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Chloride | 0.05% |
| Ignition Residue | 0.15% |
| Iron (Fe) | 30 ppm |
| Loss on Drying | 0.2% |
| Packaging | Amber Glass Bottle |
| Specific Rotation | +13.7° to +15.1° |
| Sulfate | 0.03% |
| Quantity | 1 kg |
| Percent Purity | 98.5 to 101.5% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction