Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alanyl tRNA synthetase Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Alanyl tRNA synthetase |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15713620
|
Novus Biologicals
NBP15713620UL |
20 μL |
Each for $152.22
|
|
NBP157136
|
Novus Biologicals
NBP157136 |
100 μL |
Each for $436.00
|
|
Description
Alanyl tRNA synthetase Polyclonal specifically detects Alanyl tRNA synthetase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Alanyl tRNA synthetase | |
Unconjugated | |
RUO | |
alanine tRNA ligase 1, cytoplasmic, Alanine--tRNA ligase, alanyl-tRNA synthetase, alanyl-tRNA synthetase, cytoplasmic, AlaRS, CMT2N, EC 6.1.1, EC 6.1.1.7, Renal carcinoma antigen NY-REN-42 | |
Synthetic peptides corresponding to AARS(alanyl-tRNA synthetase) The peptide sequence was selected from the C terminal of AARS. Peptide sequence VTGAEAQKALRKAESLKKCLSVMEAKVKAQTAPNKDVQREIADLGEALAT. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Polyclonal | |
Rabbit | |
P49588 | |
16 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title