Learn More
Aviva Systems Biology Corp ALAS1 Antibody - N-terminal region (ARP54346_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALAS1. Alternate names: ALAS, ALAS3, ALASH, MIG4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61181