Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALB Goat anti-Mouse, HRP, Polyclonal , Abnova™

Goat polyclonal antibody raised against mouse Alb.

Supplier:  Abnova Corporation PAB29684

Catalog No. 89-933-542

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Specifications

Specifications

ALB
Polyclonal
Goat polyclonal antibody raised against mouse Alb.
In 50mM HEPES (0.2% Ovalbumin, 0.002% Thimerosal)
Alb
Native purified mouse Alb.
1 mg
Primary
Mouse
Liquid
ELISA, Immunohistochemistry, Western Blot
HRP
ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Alb-1/Alb1
Goat
Antigen Affinity Purification
RUO
11657
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit