Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albendazole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
54965-21-8
C12H15N3O2S
265.33
HXHWSAZORRCQMX-UHFFFAOYSA-N
methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1
54965-21-8
C12H15N3O2S
265.33
HXHWSAZORRCQMX-UHFFFAOYSA-N
methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

A1411, 54965-21-8, C12H15N3O2S

Spectrum™ Albendazole, USP is a benzimidazole drug and is a broad spectrum anthelmintic that is effective against roundworms, tapeworms in humans and domestic animals. ALBENDAZOLE is on the World Health Organization′s list of essential medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 25 kg
CAS Max % 100%
Percent Purity 98 to 102%