Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Antibody (OTI3B10), HRP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270149H

Catalog No. NBP270149H


Add to Cart

Description

Description

Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Albumin
Monoclonal
HRP
PBS with No Preservative
ALB
Recombinant protein fragment corresponding to amino acids 19-609 of human Albumin (NP_000468) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot
OTI3B10
Western Blot
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
Affinity Purified
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
213.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit