Learn More
Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 5.2
For research use
Manufacturer:  Alfa Aesar™ J6552522
 View more versions of this product
Catalog No. AAJ6552522
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Lyophilized Powder
5.2
100g
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 5.2
Lyophilized powder
98%
Bovine Serum
RUO
No