Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin, Bovine, Fraction V, 96%, Standard Grade, pH 5.2, Thermo Scientific Chemicals

For research use

Supplier:  Thermo Scientific Chemicals J6585518

 View more versions of this product

Catalog No. AAJ6585518


Order in Bulk

Request bulk or custom formats

Add to Cart

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyophilized powder
50g
2° to 8°C
Albumin, Bovine, Fraction V, 96%, Standard grade, pH 5.2
Lyophilized
5.2
Bovine Serum
RUO
Native
Standard grade, 96%
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

EINECSNumber : 232-936-2

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only