Learn More
Thermo Scientific Chemicals Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0
Albumin bovine/fraction V, CAS # 9048-46-8, also known as bovine serum albumin (BSA), is a serum albumin protein derived from cows.
$60.50 - $1561.00
Specifications
Formulation | Lyophilized powder |
---|---|
pH Range | 7.0 |
Source | Bovine Serum |
Storage Requirements | 2° to 8°C |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ6465509
|
Thermo Scientific Chemicals
J6465509 |
10g |
Each for $60.50
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6465518
|
Thermo Scientific Chemicals
J6465518 |
50g |
Each
for $110.65
Save 15% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6465522
|
Thermo Scientific Chemicals
J6465522 |
100g |
Each for $214.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6465530
|
Thermo Scientific Chemicals
J6465530 |
250g |
Each for $515.50
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ64655A1
|
Thermo Scientific Chemicals
J64655A1 |
1kg |
Each for $1,561.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Albumin bovine/fraction V is a serum albumin protein commonly used in laboratory experiments including, enzyme-linked immunosorbent assay (ELISAs), immunoblot, and immune-histochemistry or -fluorescence
Applications
- Albumin bovine/fraction V displays blocking activity mainly when casein-sensitive antibodies, including phospho-specific antibodies, are used
- In genetic approaches this compound can be used to increase polymerase chain reactions (PCR) yields from low purity templates
- This compound is useful for stabilizing enzymes during storage and also for enzymatic reactions where the absence of nucleases is crucial
- In in vitro cultures, albumin bovine/fraction V has been applied as a nutrient in cell and microbial cultures especially when serum-free media is used
- It is commonly used as a protein standard to obtain calibration curves for several laboratory experiments
- Albumin bovine/fraction V is a small protein commonly used in immunohistochemistry to block nonspecific regions. The binding of this compound to nonspecific binding sites increases the probability of the antibodies bind only to the region of interest.
Specifications
Lyophilized powder | |
Bovine Serum | |
RUO | |
Native | |
Standard grade, 97% |
7.0 | |
2° to 8°C | |
Albumin, Bovine, Fraction V, 97%, Standard grade, pH 7.0 | |
Lyophilized |
Safety and Handling
EINECSNumber : 232-936-2
Recommended Storage : Keep cold
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.